Check Order Status
Lab Jacks
Actual product may vary
Fisherbrand
Lab Jacks
Precise height adjustment with the turn of a knob.
Have Questions?
Catalog Number
Height (Metric)
Height (English)
Capacity (English)
Dimensions (L x W)
FISS63080
3.81 to 13.9 cm
1.65 to 5.5 in.
31 lb.
4 x 4 in. (Plate)
Catalog number
FISS63080
Price (MYR)
-
Height (Metric):
3.81 to 13.9 cm
Height (English):
1.65 to 5.5 in.
Capacity (English):
31 lb.
Dimensions (L x W):
4 x 4 in. (Plate)
For supporting hotplates, baths, glassware and other items that require stable and precise height regulation
Large diameter knobs provide a smooth and easy height adjustment
Platforms are made of nonsparking anodized aluminum
Standard, large and giant lab jacks have threaded holes to accommodate an optional support rod (S23514) and other apparatus
Specifications
Material
Anodized Aluminum (Top), Stainless Steel
Height (Metric)
3.81 to 13.9 cm
Height (English)
1.65 to 5.5 in.
Dimensions (L x W)
4 x 4 in. (Plate)
Length (English) Base
4 in.
Width (English) Base
4 in.
Capacity (English)
31 lb.
Product Type
Lab Jack
Size
Mini
For Use With (Equipment)
Supporting hotplates, baths, glassware