Specifications
Capacity (English)2.5 gal.
Capacity (Metric)10 L
TypeUniversal Spill Kit
For Use With (Application)Safe for use on all chemicals except hydrofluoric acid
IncludesOne carton of super sorbent, six pillows, two pads, one pair each of goggles, nitrile gloves and latex boots, one disposal bag, universal instruction sheet
Unit SizeEach