Plankton Net
Plankton Net
Actual product may vary
Plankton Net
Collect minute plankton quickly and easily with this Plankton Net.
Have Questions?
Catalog NumberLength (English)
FISS45085E15 in.
Catalog number FISS45085E
Price (MYR)
-
Length (English):
15 in.
  • Take your students fishing for minute plants and animals.
  • The density of plankton in a lake or ocean can indicate the water's condition.
  • Your students can determine the density, and the nature of the plankton in water samples by using our conical plankton net.
  • Made of fine-mesh nylon, it permits the passage of water but filters out minute plankton which are collected in a 50 mL graduated tube that is fastened securely to the end of the net.
  • A stainless steel ring and harness keep the net open while it is pulled through the water.
  • The net opening is 5 in. in diameter, and the length is 15 in. from ring to tip.
Specifications
Diameter (English) Opening5 in.
Diameter (Metric) Opening12.7 cm
For Use With (Application)Collecting Plankton in Shallow Water
Product TypePlankton Net
Includes(2) 50 mL Graduated Tubes
Length (English)15 in.
MaterialNylon
Length (Metric)38.1 cm
Unit SizeEA