All Glass Vacuum Filter Holder Set and Accessories
All Glass Vacuum Filter Holder Set and Accessories
Actual product may vary
All Glass Vacuum Filter Holder Set and Accessories
All Glass Vacuum Filter Holder Set and Accessories
All Glass Vacuum Filter Holder Set and Accessories
All Glass Vacuum Filter Holder Set and Accessories used in particulate contamination analysis.
Have Questions?
Catalog NumberProduct Type
FISS43775Complete Filter Holder Set
Catalog number FISS43775
Price (MYR)
-
Product Type:
Complete Filter Holder Set
  • Used in particulate contamination analysis for filtering aqueous, organic, or corrosive liquids.
  • Recommended for HPLC solvent filtration.
  • Complete set includes a borosilicate glass funnel, anodized aluminum spring clamp, vacuum base with sintered disc, and ground joint flask.
  • Membrane filters are not included.
  • Parts are also available separately.
Specifications
For Use With (Application)Particulate Contamination Analysis
IncludesBorosilicate Glass Funnel, Anodized Aluminum Spring Clamp, Vacuum Base with Sintered Disc, and Ground Joint Flask
Item DescriptionGlass Vacuum Filter Holder
MaterialBorosilicate Glass
Product TypeComplete Filter Holder Set
Standard Taper Joint No.34/35
Unit SizeEA