Metal Draining Rack
Metal Draining Rack
Actual product may vary
Fisherbrand
Metal Draining Rack
Vinyl coated metal draining rack designed to sit on the benchtop or be hung on the wall for draining/drying of labware after washing. Includes 32 integrated pegs with rubber tips. Mounting hardware not included.
Have Questions?
Catalog NumberPositions
FISS4146732
Catalog number FISS41467
Price (MYR)
-
Positions:
32
  • Vinyl coated metal draining/drying rack
  • Hold up to 32 labware items at a time
  • Vinyl coated pegs with rubber tips are gentle on glassware
  • Can be used on the benchtop or hung on the wall
  • Mounting hardware not included
Specifications
IncludesRack with 32 pegs
Product TypeDraining Rack
Array8 x 4
AutoclavableNot Autoclavable
ColorWhite
HoldsLabware
MaterialVinyl-coated Metal
Positions32
ShapeRectangular
Shelves4 Tiers
StackableYes, When Empty
SterilityNon-sterile
For Use With (Application)Drying Labware
For Use With (Equipment)Glassware, Plasticware
MountingBenchtop or Wall
Quantity1
Length (Metric)43.8 cm
Width (Metric)30.5 cm
Height (Metric)54.6 cm
Weight (Metric)1.18 kg
Unit SizeEA