Pressure/Vacuum Pump
Pressure/Vacuum Pump
Actual product may vary
Pressure/Vacuum Pump
Features pressure and vacuum gauges.
Have Questions?
Catalog number FISS413801
Price (MYR)
-
For air sampling, pressure/vacuum filtration, degassing solutions. Separate vacuum and pressure gauges, graduated from 0–30 in.Hg and 0–100psig, respectively. Durable air seal between piston and cylinder. I
Specifications:
  • Free Air Displacement: 37L/min.
  • Pump Speed: 1725rpm
  • Pressure: 100psig (689kPag)
  • Vacuum: 25 in.hg
  • Motor: 0.17hp 110V 60Hz
  • Hose Connection takes 0.25 in. I.D. tubing
  • Dimensions: 10.75H × 15.25L × 8 in.W (27.3 × 38.7 × 20.3cm)
  • Weight: 17 lb. (7.7kg)
Specifications
Horsepower0.17 hp
IncludesMoisture trap, intake filter, handle, Rubber suction cup feet
Motor Speed1725 rpm
Electrical Requirements110 V 60 Hz
Noise Level65 to 75 dB
Weight (English)17 lb.
Weight (Metric)7.7 kg
Temperature (English) Max.104°F
Temperature (Metric) Max40°C
Temperature (Metric) Min.5°C
Flow Rate37 L/min.
Frequency60 Hz
Overall Height (English)8 in.
Overall Height (Metric)27.3 cm
Overall Length (English)10.75 in.
Overall Length (Metric)38.7 cm
Overall Width (English)15.25 in.
Maximum Pressure27 in. Hg
Voltage110 V
Unit SizeEA