Wide Mouth Glass Reagent Bottle with Glass Stopper
Wide Mouth Glass Reagent Bottle with Glass Stopper
Actual product may vary
Wide Mouth Glass Reagent Bottle with Glass Stopper
Made from high quality borosilicate 3.3 glass. Extremely durable with a low temperature gradient and superior chemical resistance.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FISS40894125 mL29/32 mm
Catalog number FISS40894
Price (MYR)
-
Capacity (Metric):
125 mL
Closure Size:
29/32 mm
  • Clear reagent bottle made from heavy duty borosilicate 3.3 glass
  • Wide mouth with interchangeable hollow borosilicate glass stopper
  • No graduations
  • Autoclavable
  • Specifications
    Capacity (Metric)125 mL
    Capacity (English)4.2 oz.
    ColorClear
    DescriptionWide neck
    For Use With (Application)Holding liquids
    IncludesInterchangeable, tight fitting, hollow glass stopper (socket size 24/29)
    MaterialBorosilicate Glass
    Length (Metric)3 cm
    Quantity1
    SterilityNon-sterile
    Weight (Metric)226.79 g
    Width (Metric)3 cm
    Closure ColorClear
    Closure IncludedYes
    AutoclavableAutoclavable
    DisposableReusable
    GraduatedNo
    MouthNarrow
    ShapeCylindrical
    Height (Metric)10.16 cm
    Product TypeReagent Bottle
    For Use With (Equipment)Liquids
    Height (English)4 in.
    Chemical LabelUnlabeled
    Closure MaterialGlass
    Closure Size29/32 mm
    Diameter (Metric)3 cm
    Closure TypeStopper
    Unit SizeEA