Acrylic Petri Dish Rack (90 mm)
Acrylic Petri Dish Rack (90 mm)
Actual product may vary
Acrylic Petri Dish Rack (90 mm)
Acrylic petri dish rack designed for use during inoculation, incubation and storage. Made with clear acrylic and white polycarbonate posts, all plates can be clearly visualized for observation during incubation. Accommodates up to sixty (60) 90 mm dia. petri dishes.
Have Questions?
Catalog NumberPositions
FISS4061060
Catalog number FISS40610
Price (MYR)
0.00
EA
Add to cart
Positions:
60
Price (MYR)
0.00
EA
Add to cart
  • Made of clear acrylic with white polycarbonate posts
  • Accommodates up to sixty (60) 90 mm sizes petri dishes.
  • Useful during inoculation, incubation and storage.
  • Visibility from all sides enables the culture to be seen and checked during incubation.
  • 4 holes drilled in separator wall for easy carrying
Specifications
Includesbase, posts
Product TypePetri Dish Rack
Array6 x 10
AutoclavableNot Autoclavable
ColorClear/White
HoldsPlates
MaterialAcrylic, Polycarbonate
Positions60
ShapeRectangular
StackableYes, When Empty
SterilityNon-sterile
For Use With (Application)Culturing
For Use With (Equipment)petri dishes
Quantity10 Pk.
Length (Metric)28.25 cm
Width (Metric)20.3 cm
Height (Metric)215.9 mm
Diameter (Metric) Inner90 mm
Weight (Metric)1.13 kg
Unit SizeEA