Improve sterilization processes with temperature indicators. The Kartell Dry Heat Sterility Indicator Tape changes color to indicate products have attained temperatures for dry heat sterilization.
Have Questions?
Catalog Number
Length (Metric)
FISS37250
50 m
Catalog number FISS37250
Price (MYR)
-
Length (Metric):
50 m
For dry heat sterilization
After 30 minutes at 155°C, the light green V stamp changes color to brown