Petri Dishes with Clear Lid
Petri Dishes with Clear Lid
Actual product may vary
Petri Dishes with Clear Lid
Feature a new and improved design and packaging.
Have Questions?
Catalog NumberDiameter (Metric)
FISS33580A100 mm
Catalog number FISS33580A
Price (MYR)
-
Diameter (Metric):
100 mm
  • Made from medical-grade, gamma sterilized polystyrene
  • Ventilation ribs on the lid allow for air circulation
  • Manufactured in a state-of-the-art cleanroom facility
  • Packaging features a UPS™ shippable outer carton to prevent damage
  • Product sleeve is made from a strong polymer to prevent bag holes
  • Dishes packed in easy-open product sleeve with tear notch
  • Product sleeves are vacuum sealed to prevent scuffing and other damage to dishes
Specifications
Closure ColorClear
Closure IncludedYes
Closure TypeStackable
ColorClear
MaterialPolystyrene
ShapeRound
SterilitySterile
TypePetri Dish
EdgesRaised Ridge
No. per Case25/Sleeve, 20 Sleeves/Cs., 500 Dishes/Cs.
Dimensions (L x W)100 x 15 mm
Height (Metric)15 mm
Depth (Metric)100 mm
Diameter (Metric)100 mm
Unit SizeBX