Plankton Net with Collection Vial
Plankton Net with Collection Vial
Actual product may vary
Plankton Net with Collection Vial
Eisco™ Plankton Net with Collection Vial features a collection bag made of strong bolting netting cloth suspended by muslin reinforcement and mounted on a wire ring.
Have Questions?
Catalog NumberGrade
FISS29535High School
Catalog number FISS29535
Price (MYR)
-
Grade:
High School
  • 35 in. deep plankton net
  • 12 in. diameter metal ring
  • Made of strong muslin netting cloth
  • 3 braided leads provide maximum strength during collection
  • Strong cloth pocket holds collection vial
  • Includes durable plastic collection vial
  • Collection vial is 2 in. deep with a 1.5 in. opening
Specifications
DescriptionBolting netting, muslin, wire
For Use With (Equipment)Insects
Includes2 collection bottles
Length (English)12 in.
Length (Metric)31 cm
MaterialMuslin netting cloth
Weight (English)0.4 lb.
Weight (Metric)181.43 g
Width (English)12 in.
Width (Metric)304.8 mm
GradeHigh School
Product TypePlankton Net
For Use With (Application)Specimen collection
Quantity1
Unit SizeEA