Class A Glass Burettes with Glass Key Stopcock
Class A Glass Burettes with Glass Key Stopcock
Actual product may vary
Fisherbrand
Class A Glass Burettes with Glass Key Stopcock
Eisco™ Class A Glass Burettes with Glass Key Stopcock designed with blue graduations and a straight single bore.
Have Questions?
Catalog NumberCapacity (Metric)
FISS2939425 mL
Catalog number FISS29394
Price (MYR)
-
Capacity (Metric):
25 mL
  • Made from high quality 3.3 borosilicate glass
  • Blue ceramic graduations
  • Straight single bore
  • Includes smooth glass key stopcock
  • Glass Key stopcock requires lubrication but offers the smoothest, most authentic control
Specifications
Capacity (Metric)25 mL
Length (Metric)1.3 cm
MaterialBorosilicate Glass
Graduations0.1 mL
For Use With (Application)Dispensing Liquid
Height (Metric)57 cm
StopcockStraight
Stopcock MaterialGlass
Tolerance±0.05 mL
TypeStraight Bore Buret
ClassClass A
Unit SizeEA