Check Order Status
PTFE Sampler/Ladle
Actual product may vary
Fisherbrand
PTFE Sampler/Ladle
Made from pure PTFE.
Have Questions?
Catalog Number
Length (Metric) Handle
FISS29080
15.2 cm
Catalog number
FISS29080
Price (MYR)
-
Length (Metric) Handle:
15.2 cm
Totally inert
With a fixed, 6 in. long (15.2 cm) handle
Reusable
Specifications
Material
PTFE
Disposable
Reusable
Color
White
Length (English) Handle
6 in.
Length (Metric) Handle
15.2 cm