Amber Reagent Bottle , Narrow Mouth with Acid Proof Polypropylene Stopper
Amber Reagent Bottle , Narrow Mouth with Acid Proof Polypropylene Stopper
Actual product may vary
Fisherbrand
Amber Reagent Bottle , Narrow Mouth with Acid Proof Polypropylene Stopper
Narrow Mouth with Acid Proof Polypropylene Stopper. Made of high quality borosilicate glass.
Have Questions?
Catalog NumberCapacity (Metric)
FISS279781000 mL
Catalog number FISS27978
Price (MYR)
-
Capacity (Metric):
1000 mL
Specifications
Capacity1000 mL
Capacity (Metric)1000 mL
Capacity (English)33.8 oz.
ColorAmber
DescriptionSocket Size: 29/32
For Use With (Application)To contain chemicals in liquid or powder form
MaterialBorosilicate Glass
Length (Metric)228.6 mm
Quantity1
SterilityNon-sterile
Standard Taper Joint29/32
Weight (Metric)453.59 g
Width (Metric)177.8 mm
Closure ColorWhite
Closure IncludedYes
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
MouthNarrow
ShapeCylindrical
Height (Metric)12.7 cm
Product TypeReagent Bottle
For Use With (Equipment)Acid
Height (English)5 in.
Chemical LabelUnlabeled
Closure MaterialPolypropylene
Closure Size29/32
Closure TypeStopper
Unit SizeEA