Check Order Status
Cross-Shaped Magnetic Stir Bars
Actual product may vary
Cross-Shaped Magnetic Stir Bars
High quality magnets coated with PTFE
Have Questions?
Catalog Number
Length (Metric)
FISS27833
38 mm
Catalog number
FISS27833
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
38 mm
High-quality magnets coated with durable PTFE
Cross shape increases turbulence and improves stirring action
Available in 2 sizes
Specifications
Autoclavable
Autoclavable
Color
White
Dimensions (L x W x H)
38 x 38 x 12 mm
Coating
PTFE
Diameter (Metric)
38 mm
Length (Metric)
38 mm
Material
PTFE, Magnet
Quantity
1/Pk.
Shape
Cross
Type
Magnetic Stir Bar