Flint Glass Bottles with Dropper
Flint Glass Bottles with Dropper
Actual product may vary
Fisherbrand
Flint Glass Bottles with Dropper
Flint Glass Bottles with Dropper designed for general use. Screw cap included with dropper assembly.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISS2776930 mL
FISS2776815 mL
Catalog number FISS27769
Price (MYR)
-
Capacity (Metric):
30 mL
  • Square Flint Glass Gottles.
  • Include screw cap with dropper assembly.
  • Non-porous glass allows for easy cleaning or autoclaving.
  • For general use.
Specifications
Capacity30 mL (1 oz.)
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorClear
For Use With (Application)Liquids or Solutions
MaterialFlint Glass
Quantity12 Pk.
SterilityNon-sterile
Closure IncludedYes
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
ShapeSquare
Height (Metric)11.4 cm
Product TypeDropping Bottle
Width (English)3 in.
Height (English)4.5 in.
Chemical LabelUnlabeled
Closure MaterialPlastic
Closure TypeDropping Bottle Cap
Through the Closure DispensingNo
Unit SizePK