Teflon Coated Laboratory Spoon Spatula
Teflon Coated Laboratory Spoon Spatula
Actual product may vary
Teflon Coated Laboratory Spoon Spatula
A universal tool for the lab
Have Questions?
Catalog NumberLength (English)
FISS270856 in.
Catalog number FISS27085
Price (MYR)
51.00
EA
Add to cart
Length (English):
6 in.
Price (MYR)
51.00
EA
Add to cart
  • Features one flat end and one spoon end
  • Stainless steel, coated in Teflon
  • Designed for use in professional laboratories
  • Also an ideal choice for science classrooms and household use
Specifications
Quantity1
AutoclavableAutoclavable
ShapeSpoon End, Spatula End
ColorSilver
Product TypeFlat End/Spoon Ended Spatula
Length (English)6 in.
Width (English)0.75 in. (Flat End), 0.80 in. (Spoon End)
MaterialTeflon Coated Stainless Steel
Length (Metric)15.25 cm
Unit SizeEA