Seamless Cellulose Dialysis Tubing
Seamless Cellulose Dialysis Tubing
Actual product may vary
Fisherbrand
Seamless Cellulose Dialysis Tubing
Retains materials with 12,000 and higher molecular weight.
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)
FISS25645C47.7 mm3 m
Catalog number FISS25645C
Price (MYR)
-
Diameter (Metric):
47.7 mm
Length (Metric):
3 m
  • Especially useful in analytical chemistry, biology, and pharmaceutical fields
  • Highly permeable to water and low molecular weight compounds
  • Contains glycerin, water and 1% sulfur
  • Must be stored in airtight container or rehumidified before use
  • Pore diameter: 4.8nm
Specifications
Product TypeDialysis Tubing
MaterialSeamless Cellulose
Diameter (Metric)47.7 mm
Diameter (English)1.88 in.
Length (English)10 ft.
Length (Metric)3 m
FormulationGlycerin, water and 1% sulfur
QuantityEach
For Use With (Application)Useful in analytical chemistry, biology, and pharmaceutical fields
Unit SizeEA