Lab Grade Sand
Lab Grade Sand
Actual product may vary
Fisherbrand
Lab Grade Sand
Fine sand for playground, laboratory chemicals.
Have Questions?
Catalog NumberQuantity
FISS25516B1 kg
Catalog number FISS25516B
Price (MYR)
-
Quantity:
1 kg
  • Fine
  • White
  • 1 kg
Specifications
PackagingPowder
Quantity1 kg
For Use With (Application)For playground, laboratory chemicals
Product TypeLab Grade Sand
Includes2.2 lb. (1kg)
Certifications/ComplianceOSHA