Check Order Status
Ticker Tape Timer Kit
Actual product may vary
Ticker Tape Timer Kit
Battery-operated, for use when no external power supply is available.
Have Questions?
Catalog number
FISS24883
Price (MYR)
-
With holder
Battery operated
Includes 12V DC timer, two rolls of tape and carbon disc
Specifications
Product Type
Ticker Tape Timer
For Use With (Application)
Measuring velocity and acceleration
Includes
Ticker timer tape (2 rolls), Carbon discs (1 Pack), Tape holder, Battery board that holds 8 D cell batteries