Boss Head Clamp, Die Cast Alloy, Powder Coated Finish, up to 16mm Rods
Boss Head Clamp, Die Cast Alloy, Powder Coated Finish, up to 16mm Rods
Actual product may vary
Fisher Catalogue
Boss Head Clamp, Die Cast Alloy, Powder Coated Finish, up to 16mm Rods
For use with laboratory stands, this popular boss head is made of die casted alloy with a powder coat finish. It accepts rods up to 16mm in diameter. Metal thumbscrews allow for adjustment. Measures approx 11x3x3cm.
Have Questions?
Catalog number FISS24263
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Specifications
TypeClamp
MaterialMetal
ColorGray
Jaw TypeFixed
Unit SizeEA