Metal ware Set
Metal ware Set
Actual product may vary
Metal ware Set
Metal ware Set
Metal ware Set
Metal ware Set
Metal ware Set
Fisherbrand
Metal ware Set
This cast iron metal ware set consists of: 1 cast base (25x26cm), 1 rod, 1 boss head clamp, 1 3-prong dual adjusting clamp, 1 cast iron ring clamp (50mmdia) and 1 cast iron ring clamp (100mm dia).
Have Questions?
Catalog NumberHeight (Metric)
FISS2371225.4 mm
Catalog number FISS23712
Price (MYR)
-
Height (Metric):
25.4 mm
Includes:
  • 1 cast base (25 x 26 cm)
  • 1 rod
  • 1 boss head clamp
  • 1 3-prong dual adjusting clamp
  • 1 cast iron ring clamp (50 mm dia.)
  • 1 cast iron ring clamp (100 mm dia)
Specifications
MaterialMetal
Height (Metric)25.4 mm