pH Paper Strips
pH Paper Strips
Actual product may vary
Fisherbrand
pH Paper Strips
An easy choice for classroom experiments or demonstrations
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpH RangeType
FISS225128 to 9.5pH Test Paper
FISS225080 to 14pH Test Paper
Catalog number FISS22512
Price (MYR)
-
pH Range:
8 to 9.5
Type:
pH Test Paper
  • Ideal for educational use
  • Low-cost paper strips are easy to pass around the classroom
  • Easy-to-usedash (em)simply dip strip in solution to be tested and match to color chart immediately
Specifications
pH Range8 to 9.5
For Use With (Application)For classroom experiments or demonstrations
Quantity100 Tests/Vial, 10 Vials/Cs.
TypepH Test Paper
Unit SizeCS