Porcelain Crucible with Gooch Filter and Cover
Porcelain Crucible with Gooch Filter and Cover
Actual product may vary
Porcelain Crucible with Gooch Filter and Cover
These Gooch crucibles offer laboratory grade porcelainware at substantial cost savings.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) OuterHeight (Metric)
FISS139652140 mL55 mm83 mm
Catalog number FISS139652
Price (MYR)
-
Capacity (Metric):
140 mL
Diameter (Metric) Outer:
55 mm
Height (Metric):
83 mm
  • Gooch crucibles are ideal for the determination of total suspended solids
  • Covers are included
  • High resistance to acids and alkalis, not including hydrofluoric acid
  • Autoclavable and glazed inside and out, except for outside bottom and rim
  • Designed to withstand temperatures up to 1150°C
  • To prevent thermal stress cracks, temperature change rate should no exceed 200°C per hour
Specifications
MaterialPorcelain
Capacity (Metric)140 mL
AutoclavableAutoclavable
Diameter (Metric) Outer55 mm
Height (Metric)83 mm
Product TypeCrucible
ShapeCylinder
Unit SizeEA