Burner Bunsen Meker: High Temperature
Burner Bunsen Meker: High Temperature
Actual product may vary
Burner Bunsen Meker: High Temperature
Burner Bunsen Meker: High Temperature is ideal for lab work requiring a hotter flame than that attainable with a bunsen burner.
Have Questions?
Catalog NumberGas Type
FISS13939LPG
Catalog number FISS13939
Price (MYR)
-
Gas Type:
LPG
  • Features adjustable orifice for natural or cylinder gas
  • Adjust the air supply with the shutter sleeve covering the air intake ports, needle valve controls gas flow
  • An expanded Meker type top with Monel stabilizing and heat intensifying grid provides a short hot flame
  • For 8 mm diameter tubing
  • Grid top diameter approximately 42 mm
Specifications
Product TypeHigh Temperature Meker Burner
For Use With (Equipment)For use with LPG (liquid petroleum gas) or butane gas
Gas TypeLPG
Length (English)8.6 in
Length (Metric)21.8 cm
MaterialStainless Steel
Diameter (English)1.7 in.
Diameter (Metric)42 mm
Diameter (Metric) Outer Mixing Tube8 mm
For Use With (Application)Lab Work Requiring a Hotter Flame than that Attainable with a Bunsen Burner
Height (English)8.5 in.
Height (Metric)215 mm
Weight (English)15 oz.
Weight (Metric)425 g
Unit SizeEA