Premium Three Finger Clamp
Premium Three Finger Clamp
Actual product may vary
Fisherbrand
Premium Three Finger Clamp
Choose durable lab equipment that will last for years. The EISCO Premium Three Finger Clamp is an industrial clamp with rubber coated fingers.
Have Questions?
Catalog NumberColor
FISS13901Blue
Catalog number FISS13901
Price (MYR)
-
Color:
Blue
  • Clamp constructed of powder coated steel
  • Maximum clamp opening is 8.5cm
  • 14cm connecting rod
  • Completely metal constructions, thread fitted actuating joints
  • 10mm diameter connecting rod
Specifications
AutoclavableAutoclavable
Type3-Prong Clamp
MaterialZinc Die Cast, Plastic Coated
Height (Metric)3.30 cm
Height (English)1.3 in.
Length (English)11 in.
Length (Metric)27.94 cm
Width (English)4 in.
Width (Metric)10.16 cm
ColorBlue
Jaw Type3-Prong
CoatingRubber
Unit SizeEA