Alcohol Lamp
Alcohol Lamp
Actual product may vary
Fisherbrand
Alcohol Lamp
Alcohol Lamp polyhedral with metal cap.
Have Questions?
Catalog number FISS13475
Price (MYR)
-
  • Allows for multiple angles
  • Capacity: 120 mL
  • Weight: 181 g (0.4 lb.)
Specifications
Product TypeAlcohol Lamp
For Use With (Equipment)Standard Molecular Biology, Sterilization Procedures
Gas TypeEthyl Alcohol
Length (English)3.8 in.
Length (Metric)9.65 cm
Length (Metric) Blade2 cm
MaterialGlass
SizeSmall
For Use With (Application)Used to produce an open flame
Height (English)2.8 in.
Height (Metric)7.11 cm
Weight (English)0.4 lb.
Weight (Metric)181 g
Width (English)2.8 in.
Width (Metric)7.11 cm
Unit SizeEA