Polypropylene Micro Pestle
Polypropylene Micro Pestle
Actual product may vary
Fisherbrand
Polypropylene Micro Pestle
Polypropylene Micro Pestle ideal for resuspending bacterial cells and precipitates of nucleic acids or proteins, and are useful for tissue grinding.
Have Questions?
Catalog number FISS13091
Price (MYR)
-
  • Double-ended with one end fits 1.5 mL microcentrifuge tubes and another end that fits 0.5 mL tubes
  • Autoclavable
Specifications
Product TypeMicro Pestle
Voltage0/1 V
MaterialPolypropylene
For Use With (Application)Resuspending bacterial cells and precipitates of nucleic acids or proteins, and are useful for tissue grinding
Holds1.5 mL and 0.5 mL Tubes
Packaging Quantity12 Pk.
Tubing StyleDouble-ended
Unit SizePK