Right-to-Know Safety-Vented Wash Bottles, GHS Labeling
Right-to-Know Safety-Vented Wash Bottles, GHS Labeling
Actual product may vary
Fisherbrand
Right-to-Know Safety-Vented Wash Bottles, GHS Labeling
Specially designed valve prevents pressure build-up, which eliminates potentially dangerous chemical drips. Allows dispensing of wash bottle in upright or inverted position.
Have Questions?
Catalog NumberCapacity (Metric)Chemical Label
FISS05129500 mLDeionized Water
Catalog number FISS05129
Price (MYR)
-
Capacity (Metric):
500 mL
Chemical Label:
Deionized Water
  • Wide mouth 53 mm (2 in.) for easy refilling
  • Translucent LDPE bottles with color-coded caps for easy identification
  • Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
  • Methyl Ethyl Ketone and Toluene do not include vented valve
  • Assortment is 4 per bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.
Specifications
Capacity500 mL, 16 oz.
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorBlue
MaterialLDPE
Quantity4/Pk.
SterilityNon-sterile
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
GraduatedNo
MouthWide, Threaded
ShapeCylindrical
Product TypeSafety-vented Wash Bottle
Chemical LabelDeionized Water
Unit SizePK