Check Order Status
Spatula Set
Actual product may vary
Fisherbrand
Spatula Set
This Spatula Set includes six of the most popular spoons and spatulas for the laboratory or the classroom.
Have Questions?
Catalog Number
Quantity
FISS04546
6/Set
Catalog number
FISS04546
Price (MYR)
-
Quantity:
6/Set
One each:
4 in. (10.2 cm) blade mounted on a hardwood handle
Microspoon with plastic handle
6 in. (15.2 cm) stainless-steel, one end flat, one end spoon
6 in. (15.2 cm) stainless-steel, one end bent, one end flat
6 in. (15.2 cm) stainless-steel, one end flat, one end tapered
6 in. (15.2 cm) polypropylene, one end spatula, one end spoon
Specifications
Quantity
6/Set
Autoclavable
Autoclavable
Shape
Flat End
Product Type
Spatula Set
For Use With (Application)
Laboratory or Classroom Use
Material
Stainless Steel