Stainless Steel Micro Spoon and Spatula, 6 in.
Stainless Steel Micro Spoon and Spatula, 6 in.
Actual product may vary
Stainless Steel Micro Spoon and Spatula, 6 in.
Stainless Steel Micro Spoon and Spatula, 6 in.
Stainless Steel Micro Spoon and Spatula, 6 in.
High-quality polished stainless steel laboratory tool with a micro spoon and spatula. Measuring 6 inches, it's durable, easy to clean, and versatile for professional labs, classrooms, kitchens, and household use. Ideal for scooping, transferring, and mixing powders and pastes.
Have Questions?
Catalog NumberLength (Metric)
FISS0309415.24 cm
Catalog number FISS03094
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
15.24 cm
  • Durable Construction: Made from high-quality polished stainless steel, ensuring longevity and easy cleaning.
  • Versatile Design: Features a dual-function tool with a micro spoon on one end and a spatula on the other, perfect for various applications.
  • Efficient Handling: Measures 6 inches in total length, excellent for scooping, transferring, and mixing powders and pastes.
  • Multi-Purpose Use: Designed for professional laboratory use, but also ideal for science classrooms, household use, restaurants, and kitchens.
This high-quality laboratory tool features a polished, stainless steel construction for durability and ease of cleaning. Measuring 6 inches in total length, this dual-function tool combines a micro spoon on one end and a spatula on the other, making it versatile for various applications. While designed for professional laboratory use, its robust and practical design makes it equally suitable for science classrooms and household use. Ideal for restaurants and kitchens, this micro spoon/spatula is excellent for scooping, transferring, and mixing powders and pastes.
Specifications
AutoclavableAutoclavable
ShapeSpoon End, Spatula End
Product TypeMicro Spoon and Spatula
Length (English)6 in.
MaterialStainless Steel
Length (Metric)15.24 cm
Unit SizeEA