Check Order Status
Steel Rods
Actual product may vary
Fisherbrand
Steel Rods
Choice of rods for all stand bases.
Have Questions?
Catalog Number
Diameter (English) Outer
Length (English)
FISS02615
0.43 in.
23.8 in.
Catalog number
FISS02615
Price (MYR)
-
Diameter (English) Outer:
0.43 in.
Length (English):
23.8 in.
Steel rod is perfect for most stand bases
Suitable for all lab applications
Choice of sizes available
Specifications
Material
Steel
Height (Metric)
60 cm
Height (English)
1.5 in.
Length (English)
23.8 in.
Length (Metric) Rod
60 cm
Diameter (English) Outer
0.43 in.
Diameter (Metric) Outer
1.1 cm
Product Type
Rod
Thread Size
0.39 x 0.05 in.
Weight (English)
11.6 lb.
Weight (Metric)
5.2 kg