Check Order Status
Base Retort Stand
Actual product may vary
Fisherbrand
Base Retort Stand
Rectangular in shape.
Have Questions?
Catalog Number
Height (English)
Length (English) Base
Weight (English)
Width (English) Base
FISS02611
1.1 in.
7.8 in.
3.6 lb.
4.9 in.
Catalog number
FISS02611
Price (MYR)
-
Height (English):
1.1 in.
Length (English) Base:
7.8 in.
Weight (English):
3.6 lb.
Width (English) Base:
4.9 in.
Pressed steel base
With hole 10 x1.5mm metric thread in the center of one short side
Finished in blue/grey colour
Specifications
Material
Steel
Height (Metric)
3 cm
Height (English)
1.1 in.
Length (English) Base
7.8 in.
Length (Metric) Base
20 cm
Width (English) Base
4.9 in.
Width (Metric) Base
12.5 cm
Product Type
Base Retort Stand
Shape
Rectangular
Thread Size
0.39 x 0.05 in.
Color
Blue/Gray
Quantity
Each
Weight (English)
3.6 lb.
Weight (Metric)
1.6 kg