Kartell™ Stackable Deep Trays
Kartell™ Stackable Deep Trays
Actual product may vary
Fisherbrand
Kartell™ Stackable Deep Trays
HDPE, stackable and low form trays.
Have Questions?
Catalog NumberCapacity (Metric)
FISS01981B20 L
Catalog number FISS01981B
Price (MYR)
-
Capacity (Metric):
20 L
Deep Trays
  • With handles for transporting and managing spills
Deep Draining Tray, Low Form
  • Rectangular tank with draining ports on all sides
  • Useful for holding defrosting samples and plant cultures
Specifications
Capacity (Metric)20 L
Dimensions (L x W x H)16.06 x 18.87 x 5.88 in. (40.7 x 47.7 x 14.93 cm)
ColorTranslucent
Height (Metric)14.93 cm
Length (Metric)40.7 cm
MaterialHDPE
TypeDeep Tray
Width (Metric)47.7 cm
IncludesHandle
Length (English)16.1 in.
Height (English)5.6 in.
ShapeRectangular
Width (English)18 in.