Class A Automatic Burette, Individually Certified
Class A Automatic Burette, Individually Certified
Actual product may vary
Class A Automatic Burette, Individually Certified
Burettes offering an affordable alternative for laboratory use.
Have Questions?
Catalog NumberCapacity (Metric)
FISS0076350 mL
Catalog number FISS00763
Price (MYR)
-
Capacity (Metric):
50 mL
  • Self-zeroing burette mounts on a glass bottle
  • Includes a PTFE needle valve stopcock, intermediate stopcock and rubber bellows
  • Class A, ASTM E-287, meets USP standards, individually serialized and certified
  • Includes Schellbach backing for easy reading of liquid levels
  • With amber graduations
  • Manufactured from precision bore tubing and calibrated To Deliver (TD, Ex) on computer-controlled machines
  • Glass bottles with ground glass joints are sold separately
Warning: Cancer and reproductive harm
Specifications
ColorClear
Capacity (Metric)50 mL
MaterialGlass
TypeAutomatic Buret
Certifications/ComplianceClass A, ASTM E-287 Individually serialized
Unit SizeEA