Kimble™ Kontes™ Microcaps™ TLC Spotting Capillaries
Kimble™ Kontes™ Microcaps™ TLC Spotting Capillaries
Actual product may vary
Fisherbrand
Kimble™ Kontes™ Microcaps™ TLC Spotting Capillaries
For spotting preadsorbent TLC plates
Have Questions?
Catalog NumberVolume (Metric)
FISK764500-00055 μL
Catalog number FISK764500-0005
Price (MYR)
627.00
CS
Add to cart
Volume (Metric):
5 μL
Price (MYR)
627.00
CS
Add to cart
Precision-bore glass capillary tubes, cut to predetermined lengths, so that each capillary tube will hold a known volume of fluid when filled.
  • Capillary action draws fluid into tube
  • Dispense fluid by squeezing bulb
  • No unusual skills or training required
  • Immediate, professional results
  • Supplied in packages containing one bulb assembly and one dispenser vial with 100 micropipets
  • Volumetric tolerance: ±1%
Specifications
Certifications/ComplianceUSP type I, ASTM E438
Diameter (English) Inner0.0174 in.
Diameter (English) Outer0.0355 in.
Diameter (Metric) Inner0.4419 mm
Diameter (Metric) Outer0.9017 mm
MaterialBorosilicate Glass
Includes1 Bulb assembly and 1 dispenser vial
Length (English)1.25 in.
Packaging TypeCase of 100 glass tubes
Volume (Metric)5 μL
Length (Metric)32 mm
TypeMicrocaps™ TLC Spotting Capillary