Kimble™ Kontes™ Pellet Pestle™
Kimble™ Kontes™ Pellet Pestle™
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Pellet Pestle™
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Have Questions?
Catalog NumberCapacity (Metric)MaterialIncludesDnase Free
FISK749520-00001.5 mLPolypropylene100 MicrotubesNo
Catalog number FISK749520-0000
Price (MYR)
1,790.00
CS
Add to cart
Capacity (Metric):
1.5 mL
Material:
Polypropylene
Includes:
100 Microtubes
Dnase Free:
No
Price (MYR)
1,790.00
CS
Add to cart
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Disposable Pellet Pestle
  • Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
  • Available with or without 100 microtubes
  • Microtubes have the same properties and come individually wrapped to ensure purity
  • Length of pestle with tip, 7cm (2.75 in.)
Reusable Pellet Pestle
  • Autoclavable CTFE polymeric head with a stainless-steel shaft
  • Pestle length with tip, 6cm (2.5 in.) for 0.5mL size; 14cm (5.59 in.) for 1.5mL size
Specifications
AutoclavableAutoclavable
ColorBlue
Capacity (English)0.05 oz.
Capacity (Metric)1.5 mL
MaterialPolypropylene
Product TypePellet Pestle
DisposableSingle-use
For Use With (Application)Resuspend Protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Length (English)2.75 in.
Dnase FreeNo
Includes100 Microtubes
For Use With (Equipment)Cordless motor
Length (Metric)70mm
Capacity0.05 oz., 1.5mL
Unit SizeCS