Kimble™ Kontes™ Gas Washing Bottle
Kimble™ Kontes™ Gas Washing Bottle
Actual product may vary
Fisherbrand
Kimble™ Kontes™ Gas Washing Bottle
Tall form gas washing bottle with full-length joint with hooks contains a fritted cylinder disc for more uniform gas distribution throughout the absorbing material. Available in two porosities: extra coarse (170-220 micron with 40/50 standard taper joint) and coarse (40-60 micron with standard 29/42 taper joint).
Have Questions?
Catalog NumberCapacity (Metric)IncludesStandard Taper Joint
FISK657750-2525250 mLFull length joint with hooks, two 2 in. springs and fritted cylinder, No.2 hose connectors40/50
Catalog number FISK657750-2525
Price (MYR)
3,487.00
CS
Add to cart
Capacity (Metric):
250 mL
Includes:
Full length joint with hooks, two 2 in. springs and fritted cylinder, No.2 hose connectors
Standard Taper Joint:
40/50
Price (MYR)
3,487.00
CS
Add to cart
  • Supplied complete with hooks and two 2 in. springs
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Capacity: 250mL, Overall Height: 315mm
  • Fits Tubing 3/8 in. I.D.
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
MaterialBorosilicate Glass
TypeTall Form Gas Washing Bottle
IncludesFull length joint with hooks, two 2 in. springs and fritted cylinder, No.2 hose connectors
Pore Size170 to 220 μm
PorosityExtra Coarse
Standard Taper Joint40/50
Height (English)12.4 in.
Height (Metric)31.5 cm
For Use With (Application)Provides more uniform gas distribution throughout the absorbing material
Certifications/ComplianceUSP Type I and ASTM E438, Type I, Class A
QuantityEach
Unit SizeCS