Kimble™ Kontes™ Erlenmeyer Flasks
Kimble™ Kontes™ Erlenmeyer Flasks
Actual product may vary
Fisherbrand
Kimble™ Kontes™ Erlenmeyer Flasks
Used to measure, mix, and store liquids
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Standard Taper Joint
FISK617000-07241000 mL23 cm24/40
FISK617000-0629500 mL20 cm29/42
FISK296500-0100100 mL11.2 cm14/20
FISK617000-0429250 mL15 cm29/42
Catalog number FISK617000-0724
Price (MYR)
468.00
CS
Add to cart
Capacity (Metric):
1000 mL
Height (Metric):
23 cm
Standard Taper Joint:
24/40
Price (MYR)
468.00
CS
Add to cart
Used to measure, mix, and store liquids
  • Single neck flask
  • Has standard taperouter joint
Specifications
Capacity (Metric)1000 mL
MaterialBorosilicate Glass
Height (English)9.05 in.
Height (Metric)23 cm
Standard Taper Joint24/40
Mouth StyleNarrow
Unit SizeCS