FLEX-COLUMNS™ are economical and easy-to-use. Short columns are typically used for ion exchange chromatography, sample desalting and semi-disposable applications. Long columns are ideally suited for gel filtration and adsorption chromatography.
Have Questions?
Catalog Number
Column Capacity
Quantity
FISK4204011015
12 mL
1
Catalog number FISK4204011015
Price (MYR)
330.00
CS
Add to cart
Column Capacity:
12 mL
Quantity:
1
Price (MYR)
330.00
CS
Add to cart
Constructed of translucent polypropylene reservoirs and column outlets permanently connected to a borosilicate glass barrel, with removable polypropylene caps
20 μm porosity High Density Polyethylene (HDPE) bed supports are fixed in the column outlets
Tubing connections on the columns are male Luer locks
A wide variety of adapters, valves, and fittings are available as accessories for use as simple and versatile tubing connections
Available in 30 different sizes with standard inside diameters of 0.7, 1.0, 1.5 and 2.5 cm and lengths ranging from 4 to 170 cm
Maximum pressure is 1.0 bar (14.7 psi, 1.0 atm, 0.1 MPa)
Temperature range is 0 to 50 °C
Sterilize with 2N NaOH or 100% ethanol
autoclaving is not recommended
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Note: End fittings and bed supports are NOT replaceable on FLEX-COLUMNS