Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Height (Metric) | Standard Taper Joint |
|---|---|---|---|
| FISK296500-0100 | 100 mL | 11.2 cm | 14/20 |
| FISK617000-0629 | 500 mL | 20 cm | 29/42 |
| FISK617000-0724 | 1000 mL | 23 cm | 24/40 |
| FISK617000-0429 | 250 mL | 15 cm | 29/42 |