Elite™ Adjustable-Volume Pipettes
Elite™ Adjustable-Volume Pipettes
Actual product may vary
Elite™ Adjustable-Volume Pipettes
Elite™ Adjustable-Volume Pipettes
Elite™ Adjustable-Volume Pipettes
Elite™ Adjustable-Volume Pipettes
Elite™ Adjustable-Volume Pipettes
Fisherbrand
Elite™ Adjustable-Volume Pipettes
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FISFBE100001 to 10 mL
FISFBE050000.5 to 5 mL
Catalog number FISFBE10000
Price (MYR)
2,117.00
EA
Add to cart
Volume (Metric):
1 to 10 mL
Price (MYR)
2,117.00
EA
Add to cart
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
  • Fully autoclavable — entire pipette can be steam autoclaved
  • Soft-touch tip ejection — a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle — comfortable grip and effortless tip attachment
  • Optimal fit with SureOne™ pipette tips — best pipetting performance
  • Super blow-out piston — creates 150% bigger air push than conventional piston for delivery of the micro size drops
Specifications
Accuracy0.5 to 2%
Compatible TipsFisherbrand Maxi Pipet tips, 1 to 10mL
AutoclavableAutoclavable
For Use WithFisherbrand™ Maxi Pipette Tip
Product LineElite
TypeStandard Pipette
Volume (Metric)1 to 10 mL
Finger StyleErgonomic
IncludesCalibration certificate, in-lab calibration tool
Increments0.02mL
Secure Tip AttachmentSoft touch tip ejection
Warranty3 Years
Unit SizeEA