Porcelain Crucible Covers
Porcelain Crucible Covers
Actual product may vary
Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) OuterDiameter (Metric) InnerHeight (Metric)Thickness (Metric)
FISFB965Z115mm105mm9mm3mm
FISFB965T44mm38mm6mm1.9mm
FISFB965V60mm54mm7mm2.5mm
Catalog number FISFB965Z
Price (MYR)
171.00
PK
Add to cart
Diameter (Metric) Outer:
115mm
Diameter (Metric) Inner:
105mm
Height (Metric):
9mm
Thickness (Metric):
3mm
Price (MYR)
171.00
PK
Add to cart
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
  • Glazed inside and out except for the outside bottom surface
  • With perforated handle
  • Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C
Specifications
DescriptionWith Perforated handle
Diameter (Metric) Inner105mm
MaterialPorcelain
FinishGlazed
For Use With (Application)Use with Fisherbrand high and wide form porcelain crucible
Max. Temperature (Metric)1150°C
Thickness (Metric)3mm
Weight (Metric)119g
Diameter (Metric) Outer115mm
Height (Metric)9mm
Product TypeCrucible Cover
Unit SizePK