Wide-Form Porcelain Crucibles
Actual product may vary
Fisherbrand
Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) OuterHeight (Metric)Weight (Metric)
FISFB965K30 mL50mm31mm26g
FKS1342971930 mL50mm31mm26g
FISFB965L50 mL61mm37mm35g
FISFB965P250 mL102mm60mm124g
FISFB965N150 mL84mm52mm90g
Catalog number FISFB965K
Price (MYR)
188.00
Pack
Capacity (Metric):
30 mL
Diameter (Metric) Outer:
50mm
Height (Metric):
31mm
Weight (Metric):
26g
Price (MYR)
188.00
Pack
Withstand temperatures up to 1150°C (2102°F)
  • Glazed inside and out except for outside bottom surface and rim
Specifications
MaterialPorcelain
FinishGlazed
Max. Temperature (Metric)1150°C
Thickness (Metric)2mm
Weight (Metric)26g
Capacity (Metric)30 mL
Diameter (Metric) Outer50mm
Height (Metric)31mm
Product TypeCrucible
Unit SizePack