Porcelain Pestles
Porcelain Pestles
Actual product may vary
Porcelain Pestles
For hand comfort in grinding
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Length (English)Length (Metric)
FISFB961O400mL Mortar7.6 in.19.4cm
FISFB961M150mL Mortar6.2 in.15.7cm
Catalog number FISFB961O
Price (MYR)
63.00
EA
Add to cart
For Use With (Equipment):
400mL Mortar
Length (English):
7.6 in.
Length (Metric):
19.4cm
Price (MYR)
63.00
EA
Add to cart
For hand comfort in grinding
  • Unglazed on grinding surface
  • Autoclavable
  • Oversized
Specifications
FinishUnglazed Grinding Surface
AutoclavableAutoclavable
Product TypePestle
MaterialPorcelain
For Use With (Equipment)400mL Mortar
For Use With (Application)For hand comfort in grinding
Length (Metric)19.4cm
Length (English)7.6 in.
Unit SizeEA