Porcelain Mortars
Porcelain Mortars
Actual product may vary
Porcelain Mortars
Glazed outside except bottom and unglazed inside
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityDiameter (English) OuterDiameter (Metric) Outer
FISFB961E13.5 oz., 400mL5.11 in.13cm
FISFB961C5.07 oz., 145mL3.54 in.9cm
Catalog number FISFB961E
Price (MYR)
92.00
EA
Add to cart
Capacity:
13.5 oz., 400mL
Diameter (English) Outer:
5.11 in.
Diameter (Metric) Outer:
13cm
Price (MYR)
92.00
EA
Add to cart
Glazed outside except bottom and unglazed inside
  • With lip
  • Deep form
  • Export quality
  • Color: white
  • Available in various capacities
Specifications
Capacity (Metric)400 mL
FinishGlazed Outer
Capacity (English)13.5 oz.
Diameter (English) Inner4.33 in.
Diameter (English) Outer5.11 in.
Diameter (Metric) Inner11cm
Diameter (Metric) Outer13cm
Product TypeMortar
MaterialPorcelain
Height (English)3.14 in.
Height (Metric)8cm
Capacity13.5 oz., 400mL
Unit SizeEA