▸
Funnels
▸
Filtering Funnels
▸
Reusable Glass Short-Stem Funnels
Actual product may vary
Fisherbrand
Reusable Glass Short-Stem Funnels
High resistance to chemical attack, mechanical, and thermal shock
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FISFB6015865
60 mL
FISFB-601-5875
90 mL
FISFB60158100
225 mL
3 Options
Catalog number
FISFB6015865
Price (MYR)
712.00
Pack
Add to cart
Capacity (Metric):
60 mL
Price (MYR)
712.00
Pack
Add to cart
High resistance to chemical attack, mechanical, and thermal shock
Constructed with heavy uniform walls, molded rims, and fire-polished stems
Formed to a 58° angle for faster, more effective filtering
Cone is suspended by its uppermost edge, leaving most of the conical area for filtration
Specifications
Diameter (Metric)Top
65 mm
Capacity (Metric)
60 mL
Diameter (Metric) Stem
8 mm
Material
Borosilicate Glass
Certifications/Compliance
ASTM E1095, Type I, Class A
Angle
58°
Diameter (Metric) Filter
110 mm
Length (Metric) Stem
63 mm