Check Order Status
Graduated Centrifuge Tube with Screw Thread Cap
Actual product may vary
Graduated Centrifuge Tube with Screw Thread Cap
Reusable large capacity 100mL tube with screw thread cap and PTFE-faced rubber liner.
Have Questions?
Catalog number
FISFB45246100
Price (MYR)
9,294.00
CS
Add to cart
Price (MYR)
9,294.00
CS
Add to cart
Standard Taper Kimax tube with a screw thread finish
Graduated and calibrated to contain
Screw-thread cap supplied has PTFE-faced rubber liner and is packaged separately
Scale, legend and marking spot are durable white ceramic enamel
Graduation Intervals mL 0-1 in 0.05, 1-3 in 0.1, 3-6 in 0.2, 6-10 in 0.5, 10-100 in 1
Tolerance 0 to 0.1 - ±0.02, Above 0.1 to 0.30 - ±0.03, Above 0.3 to 1 - ±0.05, Above 1 to 3 - ±0.10, Above 3 to 5 - ±0.20, Above 5 to 10 - ±0.50, Above 10 - ±1.00
Manufactured to the specifications found in ASTM D2158
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Product Type
Centrifuge Tube with Screw Cap