Bench Protector
Bench Protector
Actual product may vary
Bench Protector
Bench Protector
Bench Protector
Bench Protector
Fisherbrand™ Bench Protector features absorbent paper to capture spills, backed with an impermeable polyethylene to protect the covered surface from contamination
Have Questions?
Catalog NumberDimensions (L x W)Description
FISFB1200730046 x 57 cmSheet
Catalog number FISFB12007300
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
46 x 57 cm
Description:
Sheet
Features:
  • Absorbent layer of pure cellulose with smooth, firm surface
  • One side coated with impermeable polyethylene
  • Dry and liquid samples can be recovered
  • Can be incinerated for disposal
Specifications
Product TypeBench Protector
MaterialCellulose/Polyethylene
Absorbancy25 mg/cm2
Length (English)22.44 in.
Length (Metric)57 cm
Width (English)18.11 in.
Width (Metric)46 cm
Dimensions (L x W)46 x 57 cm
ColorWhite
DescriptionSheet
DisposableSingle-use
For Use With (Application)Protects benchtops, floors and carts with this highly absorbent cover
No. per Pack50
QuantityPk. of 50
Unit SizePK