Electroporation Cuvettes Plus™
Electroporation Cuvettes Plus™
Actual product may vary
Electroporation Cuvettes Plus™
Electroporation Cuvettes Plus™
Electroporation Cuvettes Plus™
Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Width (Metric) Gap
FISFB102400 μL2 mm
FISFB10190 μL1 mm
Catalog number FISFB102
Price (MYR)
-
Capacity (Metric):
400 μL
Width (Metric) Gap:
2 mm
Disposable, color-coded cuvets are compatible with most electroporators.
  • Clear, medical grade polycarbonate for high visibility
  • Polished aluminum lowers arcing frequency
  • Round cap allows single-handed removal
  • FB102 has imbedded aluminum electrodes
Specifications
SterilitySterile
TypeElectroporation Cuvet
Capacity (Metric)400 μL
MaterialPolycarbonate
IncludesImbedded aluminum electrodes
Width (Metric) Gap2 mm
For Use With (Equipment)Electroporator
Unit SizePK